Learn More
Thermo Scientific™ Remel™ TB Brilliant Green Counterstain
Catalog No. p-4525163
Change view
Click to view available options
Quantity:
5/Pk.
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R40100 Each
R40200 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R40100 Supplier Thermo Scientific™ Supplier No. R40100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel TB Brilliant Green is a counterstain for TB Kinyoun stain procedures.
Thermo Scientific™ Remel TB Brilliant Green is a counterstain for TB Kinyoun stain procedures.
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each