Learn More
Thermo Scientific™ Remel™ TB Kinyoun Carbolfuchsin Stain
Catalog No. p-4525097
Click to view available options
Quantity:
5/Pk.
Each
This item is not returnable. View return policy
This item is not returnable. View return policy
Thermo Scientific™ Remel TB Kinyoun Carbolfuchsin is a primary stain for AFB and Cryptosporidium modified acid-fast staining procedures.
Thermo Scientific™ Remel TB Kinyoun Carbolfuchsin is a primary stain for AFB and Cryptosporidium modified acid-fast staining procedures.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5/Pk.
Volume (Metric) 5 x 250 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.