Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ TB Malachite Green Counterstain
Click to view available options
Quantity:
Each
Description
Thermo Scientific™ Remel TB Malachite Green is a counterstain for TB Kinyoun stain procedure.

Specifications
Specifications
Type | TB Malachite Green |
Quantity | Each |
Color | Dark green |
pH | 2.4 - 3 |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction