Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ TB Malachite Green Counterstain

Catalog No. r40108
Click to view available options
Quantity:
Each
This item is not returnable. View return policy
This item is not returnable. View return policy

Thermo Scientific™ Remel TB Malachite Green is a counterstain for TB Kinyoun stain procedure.

Thermo Scientific™ Remel TB Malachite Green is a counterstain for TB Kinyoun stain procedure.

TRUSTED_SUSTAINABILITY

Specifications

Type TB Malachite Green
Quantity Each
Color Dark green
pH 2.4 - 3
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.