Learn More
Thermo Scientific™ Thayer Martin Agar Improved
Description
Isolate Neisseria gonorrhoeae and Neisseria meningitidis using Thayer Martin Agar Improved.
Isolate Neisseria gonorrhoeae and Neisseria meningitidis with Thermo Scientific™ Remel Thayer Martin Agar Improved. Johnston introduced a medium to grow N. gonorrhoeae in 24 hours1 and this was the advent of GC Agar Base. Thayer and Martin formulated a selective medium to isolate N. gonorrhoeae and N. meningitidis that contained antibiotics capable of supressing the commensal microbial flora present in the anal canal, vagina, pharynx and other sites2. Martin and Lewis recommended the use of anisomycin to replace nystatin and increase the shelf life of the media3.
Ordering Information:
REF R10211 and REF R10210 are Transgrow System. REF R10325 complete with bags and CO2 generating tablets.
Specifications
Specifications
Description | Thayer Martin Agar Improved |
Format | 12mm x 85mm Monoplate |
Product Type | Agar |
Quantity | 100/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.