Learn More
Thermo Scientific™ Thayer Martin Agar Modified (w/nystatin, colistin, vancomycin, trimethoprim)
Catalog No. R01881
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
15/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01881 15/Pk.
R01880 10/Pk.
R01882 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R01881 Supplier Thermo Scientific™ Supplier No. R01881
Isolate and identify Neisseria gonorrhoeae and Neisseria meningitidis with this selective, ready-to-use solid medium
Isolate Neisseria gonorrhoeae and Neisseria meningitidis with Thermo Scientific™ Remel™ Thayer Martin Agar Modified (with nystatin, colistin, vancomycin, trimethoprim)
Thayer Martin Agar is a selective medium containing antibiotics to isolate Neisseria gonorrhoeae and Neisseria meningitidis, while suppressing commensal microbial flora present in the anal canal, vagina, pharynx and other sites1
  • Nystatin inhibits Candida albicans
  • Colistin inhibits most gram-negative microorganisms, including Pseudomonas spp.
  • Vancomycin inhibits gram-positive cocci
  • Trimethoprim suppresses swarming of Proteus spp.
TRUSTED_SUSTAINABILITY
Specifications
Description Thayer Martin Agar Modified
Format 10mm x 100mm Spacesaver Plate
Product Type Agar
Quantity 15/Pk.