Learn More
Thermo Scientific™ Thayer Martin Agar Modified (w/nystatin, colistin, vancomycin, trimethoprim)
Catalog No. p-4523649
Click to view available options
Quantity:
10/Pk.
100/Pk.
15/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy
Isolate and identify Neisseria gonorrhoeae and Neisseria meningitidis with this selective, ready-to-use solid medium
Isolate Neisseria gonorrhoeae and Neisseria meningitidis with Thermo Scientific™ Remel™ Thayer Martin Agar Modified (with nystatin, colistin, vancomycin, trimethoprim)
Thayer Martin Agar is a selective medium containing antibiotics to isolate Neisseria gonorrhoeae and Neisseria meningitidis, while suppressing commensal microbial flora present in the anal canal, vagina, pharynx and other sites1
  • Nystatin inhibits Candida albicans
  • Colistin inhibits most gram-negative microorganisms, including Pseudomonas spp.
  • Vancomycin inhibits gram-positive cocci
  • Trimethoprim suppresses swarming of Proteus spp.
TRUSTED_SUSTAINABILITY
Specifications
Description Thayer Martin Agar Modified
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.