Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Thayer Martin Agar Modified (w/nystatin, colistin, vancomycin, trimethoprim)

Catalog No. r01880
Click to view available options
Quantity:
10/Pk.
100/Pk.
15/Pk.
This item is not returnable. View return policy

Catalog No. R01880

Supplier: Thermo Scientific™ R01880

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isolate and identify Neisseria gonorrhoeae and Neisseria meningitidis with this selective, ready-to-use solid medium

Isolate Neisseria gonorrhoeae and Neisseria meningitidis with Thermo Scientific™ Remel™ Thayer Martin Agar Modified (with nystatin, colistin, vancomycin, trimethoprim)

Thayer Martin Agar is a selective medium containing antibiotics to isolate Neisseria gonorrhoeae and Neisseria meningitidis, while suppressing commensal microbial flora present in the anal canal, vagina, pharynx and other sites1

  • Nystatin inhibits Candida albicans
  • Colistin inhibits most gram-negative microorganisms, including Pseudomonas spp.
  • Vancomycin inhibits gram-positive cocci
  • Trimethoprim suppresses swarming of Proteus spp.
TRUSTED_SUSTAINABILITY

Specifications

Description Thayer Martin Agar Modified
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.