Learn More
Thermo Scientific™ Thermonuclease Agar
Catalog No. p-4523101
Click to view available options
Format:
12mm x 85mm Monoplate
Presumptively identify Staphylococcus aureus with Thermonuclease Agar (w/toluidine blue).
Presumptively identify Staphylococcus aureus with Thermo Scientific™ Remel Thermonuclease Agar (w/toluidine blue). Zarzour and Belle reported the usefulness of the thermonuclease test in confirming questionable coagulase-negative S. aureus isolates1. Shanholtzer and Peterson compared the thermonuclease test with the tube coagulase and clumping factor2 and found that thermonuclease test to be as reliable as the tube coagulase and less time consuming to perform. Madison and Baselski demonstrated 100% correlation with the subsequent identification of the isolate as S. aureus by the coagulase test.
Thermonuclease Agar w/ Toluidine Blue is a solid medium recommended for use in qualitative procedures for presumptive identification of S. aureus. The nuclease produced by S. aureus is thermostable, whereas that produced by other orga
TRUSTED_SUSTAINABILITY
Specifications
Description Thermonuclease Agar w/toluidine blue
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.