Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ Todd Hewitt Broth w/Gentamicin, Nalidixic Acid, Blood
Click to view available options
Quantity:
20 x 5 mL
100 x 5 mL
Description
Todd Hewitt Broth with Gentamicin, Nalidixic Acid and Blood (Trans-Vag Broth w/5% Sheep Blood) is used for selective isolation of group B streptococci from clinical specimens containing mixed flora.
- Gentamicin and nalidixic acid inhibit some gram-negative bacteria
Specifications
Specifications
| Description | Todd Hewitt Broth; Trans-Vag Broth w/Blood; |
| Format | 15mm x 103mm Tube |
| Product Type | Microbiology Media |
| Quantity | 20 x 5 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction