Learn More
Thermo Scientific™ Tryptic Soy Agar with Lecithin, Polysorbate 80 (Microbial Content Test Agar)
Description
Detect and enumerate microorganisms present on surfaces of sanitary importance using this medium.
Detect and enumerate microorganisms present on surfaces of sanitary importance using Thermo Scientific™ Remel™ Tryptic Soy Agar with Lecithin, Polysorbate 80 (Microbial Content Test Agar). The combination of lecithin and polysorbate 80 in an agar medium effectively neutralizes disinfectants1. Tryptic Soy Agar (TSA) is prepared according to the Soybean-Casein Agar formula recommended in the United States Pharmacopeia (USP)2. TSA with Lecithin and Polysorbate 80 is compatible with environmental air sampling procedures.
Tryptic Soy Agar with Lecithin, Polysorbate 80 (Microbial Content Test Agar) is a solid medium recommended for use in qualitative procedures for the detection and enumeration of microorganisms.
Casein peptone and soy peptone provide essential amino acids, peptides and nitrogenous substances necessary for the growth of bacteria. Sodium chloride is a source of essential electrolytes and maintains osmotic equilibrium. Lecithin and polysorbate 80 neutralize residual compounds that may be present on the material being sampled. Agar is a solidifying agent.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
Description | Tryptic Soy Agar w/Lecithin, Polysorbate 80 |
Format | B Septum - 500mL |
Product Type | Agar |
Quantity | 10/Pk. |
Part Nos. R112718 and R119841 not intended for IVD use.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.