Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Tryptic Soy Agar

Catalog No. r08930
Click to view available options
Quantity:
10 x 100 mL
10 x 500 mL
10/Pk.
100 x 10 mL
100 x 22 mL
100/Pk.
20 x 22 mL
20/Pk.
Format:
15mm x 103mm Tube
15mm x 150mm Plate
20mm x 113mm Pour Tube
20mm x 113mm Tube
20mm x 150mm Pour Tube
BB - 500mL
BRB - 100mL
Monoplate
Monoplate Deep Pour
Packaging Type:
10/Pk.
100/Pk.
20/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

The Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.

Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW.
Description Tryptic Soy Agar
Format 20mm x 113mm Tube
Product Type Agar
Packaging Type 100/Pk.
Quantity 100/Pk.

REF R110092, REF R112700, and REF R112703 not intended for IVD use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.