Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Tryptic Soy Broth

Catalog No. R455056
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R455056 10 kg
R455052 500 g
R455054 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R455056 Supplier Thermo Scientific™ Supplier No. R455056
Add to Cart
Add to Cart
This item is not returnable. View return policy

Remel Tryptic Soy Broth (TSB) (Soybean Casein Digest) (Dehydrated) is used for the cultivation of a wide variety of nonfastidious microorganisms.

Remel Tryptic Soy Broth (TSB) (Soybean Casein Digest) (Dehydrated) is used for the cultivation of a wide variety of nonfastidious microorganisms.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: USP
Description Tryptic Soy Broth (TSB) (Soybean Casein Digest)
Form Powder
Product Type Dehydrated Culture Media
Quantity 10 kg

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.