Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Tryptic Soy Broth

Catalog No. p-4524494
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Remel Tryptic Soy Broth (TSB) (Soybean Casein Digest) (Dehydrated) is used for the cultivation of a wide variety of nonfastidious microorganisms.

Remel Tryptic Soy Broth (TSB) (Soybean Casein Digest) (Dehydrated) is used for the cultivation of a wide variety of nonfastidious microorganisms.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: USP
Description Tryptic Soy Broth (TSB) (Soybean Casein Digest)
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.