Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tyrosine Agar

Catalog No. r09960
Click to view available options
Quantity:
20 x 25 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Differentiate aerobic actinomycetes on the basis of tyrosine hydrolysis using Tyrosine Agar.

Differentiate aerobic actinomycetes on the basis of tyrosine hydrolysis using Thermo Scientific™ Remel™ Tyrosine Agar. Mishra et al. developed a taxonomic scheme for aerobic actinomycetes on the basis of biochemical characteristics1. A simplified scheme for identification of most commonly isolated actinomycetes was developed by McGinnis et al. and includes tyrosine, xanthine, hypoxanthine, and casein tests2. Upon extended incubation, Nocardia brasiliensis and Streptomyces spp. exhibit proteolytic activity on tyrosine agar.

The aerobic acinomycetes are a heterogeneous group of genera and include the species of Nocardia and Streptomycces3. As a group, the organisms are gram-positive, catalase positive, and may be presumptively identified by staining procedures, microscopic morphology, and substrate degradation.

  • Ready to use – Convenience of a prepared media
  • Easy to interpret – Clearing of tyrosine due to enzymatic hydrolysis around and/or under growth indicates a positive test. Growth without clearing of medium indicates a negative test.

This medium contains beef extract and peptone that provide carbohydrates, vitamins nitrogen compounds and salts essential for the growth of aerobic actinomycetes. Peptone also provides nutrients in the form of amino acids and peptides. This medium detects the presence of the hydrolytic enzyme that degrades tyrosine resulting in clearing of the media surrounding and beneath the areas of growth.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Tyrosine Agar
Format G - PT - 25mL
Product Type Agar
Quantity 20 x 25 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.