Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Urea Broth For AFB

Catalog No. R065222 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R065222 20 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R065222 Supplier Thermo Scientific™ Supplier No. R065222
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentiate Mycobacterium species based on urea hydrolysis using ready-to-use liquid medium Urea Broth For AFB.

Differentiate Mycobacterium species based on urea hydrolysis with Thermo Scientific™ Remel™ Urea Broth For AFB. In 1979, Steadham developed the urease test to help in identification of mycobacteria1. This broth test helped to achieve more consistent results for urease activity. The determination of urea hydrolysis (release of ammonia) helps in the characterization of mycobacterial strains, in particular, to distinguish scotochromogens and nonphotochromogens.2-5

Urea Broth For AFB is a liquid medium used to detect Mycobacterium species based on urea hydrolysis. This is a highly buffered medium which helps in differentiation of mycobacterial strains after 7 days incubation.

Urea Broth For AFB contains peptone and dextrose that supply essential nutrients required for growth of mycobacteria. Urea acts as the substrate for hydrolysis. Polysorbate 80 is a surfactant which aids in the dispersion of aggregates. The low pH of 5.8 facilitates a clear-cut differentiation of positive and negative results. Phenol red (1%) serves as the pH indicator.

When urea is hydrolyzed ammonia is released and the resulting alkalinity is evidenced by the phenol red indicator. A dark pink or red color development is observed in the case of a positive test whereas broth remains yellow in the case of a negative test.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Urea Broth For AFB
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 1.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.