Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ V Agar Monoplate

Catalog No. p-4523620
Click to view available options
Quantity:
10/Pk.
100/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Isolate and identify Gardnerella vaginalis with this differential, ready-to-use solid medium.

Isolate and identify Gardnerella vaginalis with Thermo Scientific™ Remel™ V Agar prepared plates, a medium containing a special blood agar base with 5% human blood, designed to enhance the growth of fastidious organisms.

Use the solid media V Agar Monoplate to isolate and identify Gardnerella vaginalis.

  • Promotes: V Agar contains the essential elements for the growth of Gardnerella vaginalis.
  • Easy to read: The transparent agar allows differentiation of G. vaginalis colonies from those of commensal microbial flora (e.g., streptococci, yeasts, lactobacilli, etc.).
  • Selective: Colistin, nalidixic acid and nystatin inhibit most gram-negative bacilli, yeast and diptheroids.
  • Ready-to-use: Convenience of prepared media.

Casein and meat peptones supply nitrogenous compounds, carbon, sulphur and trace elements necessary for the growth of G. vaginalis. Cornstarch is an energy source. Human blood is incorporated into the agar to allow for presumptive identification of G. vaginalis, which produces diffuse beta-hemolysis on V agar and Agar Selective. Colistin, nalidixic acid and nystatin are selective agents, which inhibit most gram-negative bacilli, yeast and diptheroids.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description V Agar
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.