Learn More
Labconco™ Remotely Controlled Service Fixtures
SDP
Specifications
For Use With (Equipment) Protector Hood
View More Specs
Products 4
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
16-105-30
SDP
View Documents
Labconco™
4952801
Gas Fixture N/A N/A N/A
16-105-29
SDP
View Documents
Labconco™
4952800
Standard Fixture N/A N/A N/A
16-105-31
SDP
View Documents
Labconco™
4952802
Distilled Water Fixture N/A N/A N/A
16-105-32
SDP
View Documents
Labconco™
4952803
Oxygen Fixture N/A N/A N/A
Description
Description
Protector fiber glass fume hoods can accommodate up to eight remotely controlled service fixtures, including two gooseneck faucets.
Specifications
Specifications
Protector Hood
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.