Learn More
REN Mouse anti-Human, Clone: OTI4A3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807104

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.Specifications
REN | |
Monoclonal | |
Unconjugated | |
REN | |
HNFJ2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5972 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4A3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P00797 | |
REN | |
Human recombit protein fragment corresponding to amino acids 131-406 of human REN produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.