Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ren2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112853
Description
Ren2 Polyclonal Antibody for Western Blot, ELISA
Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.
Specifications
| Ren2 | |
| Polyclonal | |
| Unconjugated | |
| Ren2 | |
| angiotensinogenase; Ren; Ren2; Ren-2; Ren-B; renin 2 tandem duplication of Ren1; renin-2; Renin-2 heavy chain; Renin-2 light chain; Rn-2; Rnr; Submandibular gland renin | |
| Rabbit | |
| Protein G | |
| RUO | |
| 19702 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3.05 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P00796 | |
| Ren2 | |
| Recombinant Mouse Renin-2 protein (64-351AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction