Learn More
Renal Cell Carcinoma Marker (gp200) Ab-1 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: PN-15
Isotype: IgG1/κ
Species Reactivity: Human and Rat. Others not known.
Epitope: Carbohydrate domain
Immunogen: Microsomal fraction of human renal cortical tissue homogenate
Molecular Weight: ∽200kDa
Positive Control: Normal kidney or renal cell CA
Cellular Localization: Membrane and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Reduced and nonreduced)
Specifications
Specifications
| Antigen | Renal Cell Carcinoma Marker (gp200) Ab-1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | PN-15 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Microsomal fraction of human renal cortical tissue homogenate |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.