Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Renal Cell Carcinoma Marker (gp200) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS409R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
12 mL
Conjugate:
Biotin
Unconjugated
Form:
Concentrated
Prediluted
Ready-to-Use
Concentration:
1mg/mL
200μg/mL
Prediluted
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate Form Concentration
MS409R1 1 mL Unconjugated Prediluted Prediluted
MS409RQ 12 mL Unconjugated Ready-to-Use Prediluted
MS409P0 100 μL Unconjugated Concentrated 200μg/mL
MS409P1ABX 100 μL Unconjugated Concentrated 1mg/mL
MS409PABX 200 μL Unconjugated Concentrated 1mg/mL
MS409B 1 mL Biotin Concentrated 200μg/mL
MS409P1 500 μL Unconjugated Concentrated 200μg/mL
MS409P 1 mL Unconjugated Concentrated 200μg/mL
MS409R7 7 mL Unconjugated Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. MS409R1 Supplier Epredia Supplier No. MS409R1

Provide accurate, reproducible results with the Epredia™ Renal Cell Carcinoma Marker (gp200) Ab-1, Mouse Monoclonal Antibody.

In normal kidney, gp200 is localized along the brush border of the pars convoluta and pars recta segments of the proximal tubule, as well as focally along the luminal surface of Bowman's capsule adjoining the outgoing proximal tubule. Of other normal tissues examined, the gp200 is also localized along the luminal surfaces of breast lobules and ducts, the luminal surface of the epididymal tubular epithelium, within the cytoplasm of parathyroid parenchymal cells, and focally within the colloid of thyroid follicles. Thirty-one other normal tissues do not express similar or cross-reacting antigens. Reportedly, gp200 is expressed by 93% of primary and 84% of metastatic renal cell carcinomas.

Host Species: Mouse

Clone: PN-15

Isotype: IgG1/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Carbohydrate domain

Immunogen: Microsomal fraction of human renal cortical tissue homogenate

Molecular Weight: ∽200kDa

Positive Control: Normal kidney or renal cell CA

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Reduced and nonreduced)


Specifications

Antigen Renal Cell Carcinoma Marker (gp200) Ab-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone PN-15
Concentration Prediluted
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Microsomal fraction of human renal cortical tissue homogenate
Quantity 1 mL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human, Rat
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.