Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Renalase Recombinant Rabbit Monoclonal Antibody (9A5T7)
GREENER_CHOICE

Catalog No. PIMA538006
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538006 Supplier Invitrogen™ Supplier No. MA538006
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Mouse liver, Mouse testis, Mouse kidney, Rat liver, Rat testis, Rat kidney Immunogen sequence: MAQVLIVGAG MTGSLCAALL RRQTSGPLYL AVWDKAEDSG GRMTTACSPH NPQCTADLGA QYITCTPHYA KKHQRFYDEL LAYGVLRPLS SPIEGMVMKE.

Renalase is a flavoprotein enzyme encoded by the RNLS gene, primarily expressed in the kidneys and heart where it plays a critical role in metabolizing catecholamines and catecholamine-like substances. It utilizes nicotinamide adenine dinucleotide (NADH) as a cofactor and operates through a superoxide-dependent mechanism, which is distinct from monoamine oxidase pathways. Renalase is considered a novel renal hormone that influences blood pressure and cardiac function by regulating circulating catecholamines. Its genetic variants have been associated with essential hypertension, highlighting its importance in cardiovascular health. In diabetic nephropathy, renalase exhibits protective effects, as demonstrated in genetically modified mouse models. These models have shown that renalase overexpression can mitigate renal damage, suggesting therapeutic potential. Renalase's expression is highest in the proximal tubules of the kidneys, which are densely innervated by sympathetic nerves, underscoring the connection between renalase and sympathetic nervous system activity. This enzyme's discovery has enriched our understanding of renal hormone regulation and its implications in diseases such as chronic kidney disease (CKD) and end-stage renal disease (ESRD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Renalase
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9A5T7
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rnls
Gene Accession No. A7RDN6, Q5U2W9
Gene Alias 6530404N21Rik; AI452315; alpha-NAD(P)H oxidase/anomerase; AW060440; C10orf59; FLJ11218; HGNC:25641; MAO-C; mMAO-C; monoamine oxidase-C; Renalase; renalase, FAD dependent amine oxidase; renalase, FAD-dependent amine oxidase; RGD1309804; RNLS
Gene Symbols Rnls
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Renalase (Q5VYX0).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361751, 67795
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.