Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Renilla Luciferase Rabbit anti-Renilla reniformis, Clone: SR07-830, Novus Biologicals™
SDP

Catalog No. NBP275644 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275644 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275644 Supplier Novus Biologicals Supplier No. NBP275644

Rabbit Monoclonal Antibody

Renilla Luciferase Monoclonal specifically detects Renilla Luciferase in Renilla reniformis samples. It is validated for Western Blot.

Specifications

Antigen Renilla Luciferase
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone SR07-830
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Host Species Rabbit
Immunogen Recombinant protein within Sea pansy Renilla Luciferase aa 1-96/ 311. (SwissProt: P27652 Renilla reniformis)
Molecular Weight of Antigen 36 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cell Biology, Cellular Markers, Epitope Tags, Fusion Proteins
Primary or Secondary Primary
Target Species Renilla
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.