Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Renilla luciferase Recombinant Rabbit Monoclonal Antibody (SR07-830)
GREENER_CHOICE

Catalog No. PIMA532013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532013 Supplier Invitrogen™ Supplier No. MA532013
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Renilla luciferase (RLuc), derived from the sea pansy Renilla reniformis, is a widely-used reporter gene in molecular biology for in vitro and in vivo assays, including gene expression studies and small animal imaging. The luciferase enzyme, approximately 37 kDa in size, belongs to the alpha/beta hydrolase family and operates via a catalytic mechanism involving the oxidative decarboxylation of its substrate, coelenterazine, in the presence of molecular oxygen, resulting in the emission of blue light. The gene encoding Renilla luciferase has been isolated and characterized, with its cDNA available for various applications. Structurally, RLuc contains a hydrolase fold akin to that of bacterial dehalogenases, suggesting an evolutionary relationship. Despite extensive utilization, the exact details of its active site and catalytic mechanism continue to be explored. The bioluminescent property of RLuc is particularly useful for generating a detectable signal in various biological assays, enhancing its utility as a biosensor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Renilla luciferase
Applications Western Blot
Classification Recombinant Monoclonal
Clone SR07-830
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% Glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene Alias Renilla-luciferin 2-monooxygenase
Host Species Rabbit
Immunogen Recombinant protein within Sea pansy Renilla Luciferase aa 1-96.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Renilla
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.