Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Renin Antibody (OTI2F10), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273851UV
Description
Renin Monoclonal specifically detects Renin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Renin | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
REN | |
Recombinant protein fragment corresponding to amino acids 131-406 of human Renin (NP_000528) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2F10 | |
Western Blot, Immunohistochemistry | |
angiotensin-forming enzyme, Angiotensinogenase, EC 3.4.23, EC 3.4.23.15, FLJ10761, HNFJ2, Prorenin, renin, renin precursor, renal | |
Mouse | |
Affinity Purified | |
Cardiovascular Biology, metabolism | |
5972.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction