Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Renin Monoclonal Antibody (3E2B11C4)
GREENER_CHOICE

Catalog No. PIMA529521
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529521 Supplier Invitrogen™ Supplier No. MA529521
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Renin/REN.

Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Renin
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3E2B11C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Ren
Gene Accession No. P00797
Gene Alias angiotensin-forming enzyme; angiotensinogenase; aspartyl-protease; D19352; EC 3.4.23; EC 3.4.23.15; FLJ10761; HNFJ2; kidney renin; RATRENAA; Ren; Ren1; Ren-1; Ren1c; Ren1d; Ren-A; RENAA; Renin; renin 1 structural; renin b; renin precursor, renal; renin-1; Rn-1; Rnr
Gene Symbols Ren
Host Species Mouse
Immunogen Recombinant Human Renin protein (Met1-Arg406).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5972
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.