Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Renin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102432
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102432 Supplier Invitrogen™ Supplier No. PA5102432
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Renin.

Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Renin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ren
Gene Accession No. P00797, P06281, P08424
Gene Alias angiotensin-forming enzyme; angiotensinogenase; aspartyl-protease; D19352; EC 3.4.23; EC 3.4.23.15; FLJ10761; HNFJ2; kidney renin; RATRENAA; Ren; Ren1; Ren-1; Ren1c; Ren1d; Ren-A; RENAA; Renin; renin 1 structural; renin b; renin precursor, renal; renin-1; Rn-1; Rnr
Gene Symbols Ren, Ren1
Host Species Rabbit
Immunogen A synthesized peptide derived from human REN(Accession P00797), corresponding to amino acid residues V206-F256.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19701, 24715, 5972
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.