Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Renin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110692
Description
Immunogen sequence: FHYINLIKTG VWQIQMKGVS VGSSTLLCED GCLALVDTGA SYISGSTSSI EKLMEALGAK KRLFDYVVKC NEGPTLPDIS FHLGGKEYTL TSADYVFQES YSSKKLCTLA IHAMDIPPPT GPTWALGATF IRKFYTEFD.
Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.
Specifications
| Renin | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P00797 | |
| Ren | |
| Recombinant Protein corresonding to Human Renin. Recombinant protein control fragment (Product #RP-101634). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Ren | |
| angiotensin-forming enzyme; angiotensinogenase; aspartyl-protease; D19352; EC 3.4.23; EC 3.4.23.15; FLJ10761; HNFJ2; kidney renin; RATRENAA; Ren; Ren1; Ren-1; Ren1c; Ren1d; Ren-A; RENAA; Renin; renin 1 structural; renin b; renin precursor, renal; renin-1; Rn-1; Rnr | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5972 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction