Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Renin R Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 9 publications
Supplier: Novus Biologicals NB1001318
Description
Renin R Polyclonal specifically detects Renin R in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
Renin R | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
APT6M8-9, ATP6M8-9MRXE, ATPase H(+)-transporting lysosomal accessory protein 2, ATPase H(+)-transporting lysosomal-interacting protein 2, ATPase, H+ transporting, lysosomal accessory protein 2, ATPase, H+ transporting, lysosomal interacting protein 2, CAPER, ELDF10, Embryonic liver differentiation factor 10, ER-localized type I transmembrane adaptor, H+ transporting, lysosomal (vacuolar proton pump) membrane sectorassociated protein M8-9, M8-9, MGC99577, MSTP009, N14F, renin receptor, vacuolar proton ATP synthase membrane sector associated protein M8-9, V-ATPase M8.9 subunit, XMRE | |
Goat | |
39 kDa | |
0.1 mg | |
Primary | |
Expected from sequence similarity: Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1.0 ug/ml, Flow Cytometry 10 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:8000 | |
O75787 | |
ATP6AP2 | |
Peptide with sequence C-SIIYRMTNQKIRMD corresponding to C-Terminus according to NP_005756.2. | |
Affinity Purified | |
RUO | |
10159 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction