Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Renin Rabbit anti-Human, Clone: 9, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | Renin |
---|---|
Clone | 9 |
Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:25-1:100, ELISA 1:25000-1:50000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:500-1:2000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Renin Monoclonal specifically detects Renin in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Renin | |
Western Blot 1:500-1:1000, Flow Cytometry 1:25-1:100, ELISA 1:25000-1:50000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
5972 | |
IgG | |
Protein A purified |
9 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
angiotensin-forming enzyme, Angiotensinogenase, EC 3.4.23, EC 3.4.23.15, FLJ10761, HNFJ2, renin, renin precursor, renal | |
Recombinant Human Renin protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title