Learn More
Invitrogen™ Renin Recombinant Rabbit Monoclonal Antibody (1)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA529766
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse REN1/Renin-1.
Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.
Specifications
Renin | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P06281 | |
Ren1 | |
Recombinant Mouse REN1 protein (Met1-Arg402). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
1 | |
Unconjugated | |
Ren1 | |
angiotensin-forming enzyme; angiotensinogenase; aspartyl-protease; D19352; EC 3.4.23; EC 3.4.23.15; FLJ10761; HNFJ2; kidney renin; RATRENAA; Ren; Ren1; Ren-1; Ren1c; Ren1d; Ren-A; RENAA; Renin; renin 1 structural; renin b; renin precursor, renal; renin-1; Rn-1; Rnr | |
Rabbit | |
Protein A | |
RUO | |
19701 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.