Learn More
RENT1 Polyclonal Antibody, Bethyl Laboratories
Goat Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300038AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein is located only in the cytoplasm. When translation ends, it interacts with the protein that is a functional homolog of yeast Upf2p to trigger mRNA decapping. Use of multiple polyadenylation sites has been noted for this gene.Specifications
| RENT1 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q92900, Q9EPU0 | |
| UPF1 | |
| Between 1075 and C-term | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| UPF1 | |
| Regulator of nonsense transcripts 1; ATP-dependent helicase RENT1; Nonsense mRNA reducing factor 1; NORF1; Up-frameshift suppressor 1 homolog; hUpf1; RENT1; KIAA0221; UPF1; | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 19704, 5976 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.