Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RENT1/UPF1/hUPF1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100370JF549
Description
RENT1/UPF1/hUPF1 Polyclonal antibody specifically detects RENT1/UPF1/hUPF1 in Human, Mouse samples. It is validated for Western BlotSpecifications
RENT1/UPF1/hUPF1 | |
Polyclonal | |
Western Blot | |
ATP-dependent helicase RENT1, EC 3.6.1, EC 3.6.4.-, FLJ43809, HUPF1, KIAA0221FLJ46894, Nonsense mRNA reducing factor 1, NORF1delta helicase, pNORF1, regulator of nonsense transcripts 1, RENT1UP Frameshift 1, UPF1 regulator of nonsense transcripts homolog (yeast), up-frameshift mutation 1 homolog, Up-frameshift suppressor 1 homolog, yeast Upf1p homolog | |
The immunogen recognized by this antibody maps to a region between residue 1075 and the C-terminus (residue 1118) of human Regulator of Nonsense Transcripts 1 (up-frameshift mutation 1 homolog) using the numbering given in entry NP_002902.2 (GeneID 5976). | |
0.1 mL | |
DNA Repair, Transcription Factors and Regulators | |
5976 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction