Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RENT1/UPF1/hUPF1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100368
Description
RENT1/UPF1/hUPF1 Polyclonal specifically detects RENT1/UPF1/hUPF1 in Human, Mouse samples. It is validated for Western Blot.Specifications
RENT1/UPF1/hUPF1 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
ATP-dependent helicase RENT1, EC 3.6.1, EC 3.6.4.-, FLJ43809, HUPF1, KIAA0221FLJ46894, Nonsense mRNA reducing factor 1, NORF1delta helicase, pNORF1, regulator of nonsense transcripts 1, RENT1UP Frameshift 1, UPF1 regulator of nonsense transcripts homolog (yeast), up-frameshift mutation 1 homolog, Up-frameshift suppressor 1 homolog, yeast Upf1p homolog | |
Goat | |
Affinity Purified | |
RUO | |
5976 | |
Store at 4C. Do not freeze. |
Western Blot | |
1.0 mg/mL | |
Western Blot 1:1 | |
NP_002902.2 | |
UPF1 | |
The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human Regulator of Nonsense Transcripts 1 (up-frameshift mutation 1 homolog) using the numbering given in entry NP_002902.2 (GeneID 5976). | |
100 μL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction