Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REP15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563785
missing translation for 'changeView'
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563785 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. PIPA563785 Supplier Invitrogen™ missing translation for 'supplierNo' PA563785
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FEYPPSKLCP AANTLNEIFL IHFITFCQEK GVDEWLTTTK MTKHQAFLFG ADWIWTFWGS DKQIKLQLAV QTLQMSSPPP VESKPCDLSN PESRV Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 67%.

The protein encoded by this intronless gene interacts with GTP-bound Rab15 and is involved in recycling of transferrin receptor from the endocytic recycling compartment to the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REP15
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene REP15
Gene Accession No. Q6BDI9
Gene Alias 2210417D09Rik; Rab15 effector protein; RAB15EP; Rep15; RGD1562899
Gene Symbols REP15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human REP15. Recombinant protein control fragment (Product #RP-103679).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387849
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.