Description
- Ideal for viscous, dense and volatile liquides
- Minimum increments printed on the syringe itself
- Accurately deliver selected dose
- Made from high quality polypropylene and polyethylene
- Pyrogen-, ATP-, RNase-, and DNase-free
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | ISO8655 ISO11137 |
| Material | Polypropylene |
| Sterility | Sterile |
| For Use With (Application) | Dispensing of viscous and volatile liquids |
| Quantity | 100/Pk. |
| Volume (Metric) | 1.25 mL |