Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Replacement Battery Covers for Label Printers

Catalog No. p-4826122 Shop All Brady Corporation Products
Click to view available options
Color:
Gray
Yellow
For Use With (Equipment):
BMP21-LAB Label Printer
BMP21-PLUS Label Printer

Catalog No. 19-102-831

Supplier: Brady™ BMP21LABBATTCOV

Only null left
Add to Cart
Add to Cart

Replacement battery covers for BMP™21 portable, BMP21-Lab portable and BMP21-Plus portable label printers

  • Plastic cover snaps in place over battery compartment

Specifications

Product Type Replacement Battery Cover
Color Gray
For Use With (Equipment) BMP21-LAB Label Printer
Material Plastic
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.