Learn More
Restek™ Replacement Chemical Traps for Agilent™ Gas Chromatographs
SDP
Catalog No. 06712139 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
O-rings for Trap Fittings
Replacement Split Vent Trap
Replacement Trap
Split Vent Trap Assembly
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-712-139 Replacement Trap
06-712-138 Split Vent Trap Assembly
06-718-767 O-rings for Trap Fittings
06-712-056 Replacement Split Vent Trap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-712-139 Supplier Restek™ Supplier No. 23032
Add to Cart
Add to Cart
Easy to install replacement chemical traps attach to the same fittings as original Agilent GC equipment
  • Built-in frits and adsorbents remove both moisture and hydrocarbons
  • Trap can be regenerated to remove contaminants and restore performance
  • Also available is a Replacement 32 in. Split Line with all installation hardware included
Specifications
Type Replacement Trap
For Use With (Application) Gas Chromatography
For Use With (Equipment) Agilent GCs