Learn More
Fisherbrand™ Replacement Deuterium Lamp For Shimadzu UV and Visible HPLC Detector
Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Shimadzu LC2010 HPLC Detector
Description
- Long life
- Drop-in OEM replacement
- Traceable by serial number
Order Info
Cat. No. 14-500-727 is comparable to Shimadzu part number 228-37401-00.
Compliance
WEEE and ROHS compliant
Specifications
Specifications
Light Source | Deuterium |
For Use With (Equipment) | Shimadzu LC2010 HPLC Detector |
Product Type | Replacement Detector Lamp |