Learn More
Fisherbrand™ Replacement Deuterium Lamp for Waters UV and Visible HPLC Detector
Catalog No. p-7075335 Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Waters 2487 HPLC Detector
Fisherbrand™ Replacement Deuterium Lamp for Waters UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
  • Long life
  • Drop-in OEM replacement
  • Traceable by serial number
Order Info
Cat. No. 14-500-731 is comparable to Waters part number WAS081142.
Compliance
WEEE and ROHS compliant
Specifications
Light Source Deuterium
For Use With (Equipment) Waters 2487 HPLC Detector
Product Type Replacement Detector Lamp
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.