Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synthware™ Replacement Double Oblique Bore Stopcock

Shop All Kemtech Products


Click to view available options
Plug Bore:
2 mm
4 mm
Specifications
Specifications
| Type | Double Oblique Stopcock |
| Plug Bore | 4 mm |
| Material | Borosilicate Glass |
| For Use With (Application) | Organic Synthesis |
| For Use With (Equipment) | Three-way Tap |
| Plug Material | PTFE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction