Learn More
Restek™ Replacement FID Collector Assembly for Agilent 6890/6850 GCs
SDP
Catalog No. p-3186927
Click to view available options
Type:
FID Collector Assembly Kit
For use with Agilent 6850, 6890, 7890 GCs.
  • Constructed of high-quality stainless steel
  • Individual replacement parts available
Specifications
Type FID Collector Assembly Kit
For Use With Agilent 6890/6850
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.