Learn More
Cole-Parmer™ Replacement Items for Freezer/Mill™ Grinding Vial Sets
Catalog No. 6801C20
Change view
Click to view available options
Product Type:
Center Cylinder
End Plug
Replacement Impactor
Material:
Polycarbonate
Stainless Steel (440 C)
For Use With (Equipment):
6761 Vial Set
6771 Vial Set
6801 Large Grinding Vial Set and 6871 Large Cr-Free Grinding Vial Set
6881 Vial Set
Spex SamplePrep Freezer/Mill Cryogenic Grinder
Packaging Quantity:
1 Ea.
2 Pk.
20 Pk.
4 Pk.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material For Use With (Equipment) Packaging Quantity
6801C20 Center Cylinder Polycarbonate 6801 Large Grinding Vial Set and 6871 Large Cr-Free Grinding Vial Set 20 Pk.
6751C4 - Polycarbonate Spex SamplePrep Freezer/Mill Cryogenic Grinder 4 Pk.
6801C4 Center Cylinder Polycarbonate 6801 Large Grinding Vial Set and 6871 Large Cr-Free Grinding Vial Set 4 Pk.
6751C20 Center Cylinder Polycarbonate Spex SamplePrep Freezer/Mill Cryogenic Grinder 20 Pk.
68-81C-20 Center Cylinder Polycarbonate 6881 Vial Set 20 Pk.
6751E End Plug Stainless Steel (440 C) Spex SamplePrep Freezer/Mill Cryogenic Grinder 2 Pk.
67-61E End Plug - 6761 Vial Set 2 Pk.
67-71E End Plug - 6771 Vial Set 2 Pk.
6751P Replacement Impactor Stainless Steel (440 C) Spex SamplePrep Freezer/Mill Cryogenic Grinder 1 Ea.
67-61P Replacement Impactor - 6761 Vial Set 1 Ea.
67-71P Replacement Impactor - 6771 Vial Set 1 Ea.
68-81P Replacement Impactor - - 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 6801C20 Supplier Cole-Parmer™ Supplier No. 6801C20
Add to Cart
Add to Cart
Keep spare items to ensure there is no down time.
  • Suitable for use with the Cole-Parmer™ Freezer/Mill™ grinding vial sets
  • Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinding vial sets with a wide range of grinding ball options suitable for your application.
Specifications
Product Type Center Cylinder
For Use With (Equipment) 6801 Large Grinding Vial Set and 6871 Large Cr-Free Grinding Vial Set
Material Polycarbonate
Model Cole-Parmer Freezer/Mill
Packaging Quantity 20 Pk.