Learn More
Cole-Parmer™ Replacement Items for Freezer/Mill™ Grinding Vial Sets
Description
Requires
886 Mid-Size Grinding Vial Adapter for grinding in the 6870 Large Freezer/Mill and 6884 Mid-Size Grinding Vial Adapter for Large Extractor/Vial Opener.
- Suitable for use with the Cole-Parmer™ Freezer/Mill™ grinding vial sets
- Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinding vial sets with a wide range of grinding ball options suitable for your application.
Specifications
Specifications
| Product Type | Center Cylinder |
| For Use With (Equipment) | 6881 Vial Set |
| Material | Polycarbonate |
| Model | Cole-Parmer Freezer/Mill |
| Packaging Quantity | 20 Pk. |