Learn More
Labconco™ Glass Manifold
SDP
Catalog No. 21130B Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Application):
Kjedahl Digestion
Catalog No. For Use With (Application)
21-130B Kjedahl Digestion
1 options
Catalog No. 21-130B Supplier Labconco™ Supplier No. 6001100
Add to Cart
Add to Cart
Make exhaustion of fumes from Kjeldahl digestions more convenient by adding a Labconco Glass Manifold to your system.
  • Six-port glass manifold exhausts fumes from Kjeldahl digestions
  • Serrated connector for water aspirator
Specifications
Product Type Fume Removal System
For Use With (Application) Kjedahl Digestion
Material Glass
WARNING: Cancer - www.P65Warnings.ca.gov