Learn More
Trajan Scientific and Medical™ Replacement Needle and Plunger Kit for Shimadzu Autosampler Syringes
SDP
Catalog No. 033738 Shop All SGE Inc. Products
Change view
Click to view available options
Needle Gauge:
23 G
26 G
Diameter (Metric) Outer:
0.47 mm
0.63 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Diameter (Metric) Outer
03-373-8 26 G 0.47 mm
03-374-5 23 G 0.63 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-373-8 Supplier Trajan Scientific and Medical™ Supplier No. 033738
Repair kit for Shimadzu Removable Needle Autosampler Syringes.
For use with Shimadzu Removable Needle Syringes for AOC14, AOC17 and AOC 20 Autosamplers
  • Includes spare needle and plunger
  • Plunger extends to needle tip
  • Sample contained in needle only
  • Zero dead volume
  • Needle and plunger must be replaced together
Specifications
Needle Gauge 26 G
Diameter (Metric) Outer 0.47 mm
Product Type Replacement Needle Kit
Needle Point Style Cone
For Use With (Equipment) Syringes (0.5μL)