Learn More
Restek™ Replacement Parts for FID Collector Housing Kits
SDP
Catalog No. 06611259 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
Type:
Collector Housing
Replacement Silicone Washers
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-611-259 Replacement Silicone Washers
06-611-256 Collector Housing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-611-259 Supplier Restek™ Supplier No. 23064
Extend the useful life of FID Collector Housing Kits
Specifications
Type Replacement Silicone Washers
For Use With (Application) Agilent 5890/6890/7890
Quantity 12/Pk.
For Use With (Equipment) FID Collector Housing Kits