Learn More
Trajan™ SilTite™ FingerTite Ferrule
SDP
Catalog No. SG073632 Shop All SGE Inc. Products
Change view
Click to view available options
Diameter (Metric) Inside:
0.4 mm
0.5 mm
0.7 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inside
SG073632 0.7 mm
SG073631 0.5 mm
SG073630 0.4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SG073632 Supplier Trajan™ Supplier No. 073632
For gas chromatography systems
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Type GC Ferrule
Quantity 10/Pk.
Diameter (Metric) Inside 0.7 mm