Learn More
Victory Innovations™ Replacement Three-In-One Nozzle for VP200ESK and VP300ES
Description
Victory Innovations™ 3-in-1 Nozzle matches chemical dwell times with 40-micron, 80-micron, and 110-micron settings.
For use with Handheld and Backpack Victory™ Sprayers
Specifications
Specifications
| For Use With (Equipment) | Electrostatic Sprayers VP200ESK and VP300ES |
| Technology | 40-micron, 80-micron, and 110-micron Settings |
| Type | Handheld Sprayer Accessories |